Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF133 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598251
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598251 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598251 Supplier Invitrogen™ Supplier No. PA598251
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF133 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

May be involved in transcriptional regulation as a repressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF133
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ZNF133
Gene Accession No. P52736
Gene Alias pHZ-13; pHZ-66; zinc finger protein 133; zinc finger protein 133 (clone pHZ-13); Zinc finger protein 150; zinc finger protein 150 (pHZ-66); ZNF133; ZNF150
Gene Symbols ZNF133
Host Species Rabbit
Immunogen Recombinant Human Zinc finger protein 133 protein (1-210AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7692
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.