Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF180 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556026
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556026 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556026 Supplier Invitrogen™ Supplier No. PA556026
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QEILLQEVAF TQRKAVIHER VCKSDETGEK SGLNSSLFSS PVIPIRNHFH KHVSHAKKWH LNAAVNSHQK INENETLYEN NECGKPPQSI HLIQFTRTQT KDKCYGFSDR IQSFCHGTPL HIHEKIHGGG KTFDFKECGQ Highest antigen sequence identity to the following orthologs: Mouse - 39%, Rat - 43%.

Zinc finger proteins have been shown to interact with nucleic acids and to have diverse functions. The zinc finger domain is a conserved amino acid sequence motif containing 2 specifically positioned cysteines and 2 histidines that are involved in coordinating zinc. Kruppel-related proteins form 1 family of zinc finger proteins. See MIM 604749 for additional information on zinc finger proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF180
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF180
Gene Accession No. Q9UJW8
Gene Alias HHZ168; zinc finger protein 180; ZNF180
Gene Symbols ZNF180
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF180. Recombinant protein control fragment (Product #RP-99725).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7733
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.