Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF187 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101179
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101179 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101179 Supplier Invitrogen™ Supplier No. PA5101179
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ZNF187.

ZNF187 may be involved in transcriptional regulation (Probable).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF187
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ZSCAN26
Gene Accession No. Q16670
Gene Alias Azf8; BC068174; Protein SRE-ZBP; SREZBP; SRE-ZBP; Zfp187; zinc finger and SCAN domain containing 26; zinc finger and SCAN domain-containing protein 26; Zinc finger protein 187; zinc finger protein 8; Znf187; Zscan26
Gene Symbols ZSCAN26
Host Species Rabbit
Immunogen A synthesized peptide derived from human ZSCAN26(Accession Q16670), corresponding to amino acid residues R119-C169.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7741
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.