Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF217 Monoclonal Antibody (OTI2B1)
GREENER_CHOICE

Catalog No. PIMA527088
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527088 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527088 Supplier Invitrogen™ Supplier No. MA527088
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZNF217 Monoclonal Antibody for Western Blot, IHC (P)

Binds to the promoters of target genes and functions as repressor. Promotes cell proliferation and antagonizes cell death. Promotes phosphorylation of AKT1 at 'Ser-473'.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF217
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2B1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ZNF217
Gene Accession No. O75362
Gene Alias ZABC1; Zinc finger protein 217; ZNF217
Gene Symbols ZNF217
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 730-1048 of ZNF217 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7764
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.