Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Metal Amalgamated, Granular, Spectrum™ Chemical
SDP

Catalog No. 18607858 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-858 100 g Poly Bottle
18-607-859 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 18-607-858 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. Z1030100GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Z1030, Zn-Hg

Spectrum™ Chemical Zinc Metal Amalgamated, Granular is zinc that has been surface treated with mercury to form a surface amalgam. It is a powerful reducing agent and because the amalgam does not have a passive layer, it is a more effective reducing agent than zinc metal. It is moderately stable in air and is a good starting material for the preparation of solutions of titanium(III), vanadium(II), chromium(II), molybdenum(III), niobium(III) and uranium(III).

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
IUPAC Name zinc
SMILES [Zn]

Specifications

Physical Form Granular/Lump
CAS Max % 1
Quantity 100 g
Packaging Poly Bottle
Grade Not Available
Chemical Name or Material Zinc Metal Amalgamated, Granular
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.