Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF253 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501734753 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4753 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4753 Supplier Proteintech Group Inc Supplier No. 251521AP150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

ZNF253 may function as a transcription factor. Seems to have a transcriptional repression activity.

Specifications

Antigen ZNF253
Applications Western Blot
Classification Polyclonal
Concentration 0.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ZNF253
Gene Accession No. O75346
Gene Alias BMZF 1, BMZF1, Bone marrow zinc finger 1, zinc finger protein 253, Zinc finger protein 411, ZNF253, ZNF411
Gene Symbols ZNF253
Host Species Rabbit
Immunogen ZNF253 Fusion Protein Ag18570
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56242
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.