Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF274 Monoclonal Antibody (1D8)
GREENER_CHOICE

Catalog No. PIMA521159 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA521159 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA521159 Supplier Invitrogen™ Supplier No. MA521159
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Peptide Sequence: QKIDNPESQA NSGALDTNQV LLHKIPPRKR LRKRDSQVKS MKHNSRVKIH QKSCERQKAK EGNGCRKTFS RSTKQITFIR IHKGSQVCRC SECGKIFRNP RYFSVHKKIH T.

ZNF274 is a zinc finger protein containing five C2H2-type zinc finger domains, one or two Kruppel-associated box A (KRAB A) domains, and a leucine-rich domain. The protein has been suggested to be a transcriptional repressor. It localizes predominantly to the nucleolus.This gene encodes a zinc finger protein containing five C2H2-type zinc finger domains, one or two Kruppel-associated box A (KRAB A) domains, and a leucine-rich domain. The encoded protein has been suggested to be a transcriptional repressor. It localizes predominantly to the nucleolus. Alternatively spliced transcript variants encoding different isoforms exist. These variants utilize alternative polyadenylation signals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF274
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1D8
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene ZNF274
Gene Accession No. Q96GC6
Gene Alias DKFZp686K08243; HFB101; KRAB zinc finger protein HFB101; neurotrophin receptor-interacting factor homolog; SP2114; Zf2; zinc finger protein 274; Zinc finger protein HFB101; zinc finger protein with KRAB and SCAN domains 19; Zinc finger protein zfp2; ZKSCAN19; ZNF274; ZSCAN51
Gene Symbols ZNF274
Host Species Mouse
Immunogen ZNF274 (NP_598009, 420 a.a. ~ 530 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10782
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.