Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF302 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120643
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120643 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120643 Supplier Invitrogen™ Supplier No. PA5120643
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: DEDSPQPVTM EKVVKQSYEF SNSNKNLEYT ECDTFRSTFH SKSTLSEPQN NSAEGNSHKY DILKKNLSKK SVIKSERING GKKLLNSNKS GAAFNQSKSL TLPQTCNREK I.

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF302
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4.73 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Alias HSD16; MST154; MSTP154; Zinc finger protein 135-like; Zinc finger protein 140-like; Zinc finger protein 302; zinc finger protein 327; ZNF135L; ZNF140L; ZNF302; ZNF327
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 90-200 of human ZNF302 (NP_060913.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.