Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF321P Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569740
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569740 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569740 Supplier Invitrogen™ Supplier No. PA569740
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF321P Polyclonal Antibody for Western Blot

ZNF321P (Zinc Finger Protein 321, Pseudogene) is a Pseudogene. Gene Ontology (GO) annotations related to this gene includenucleic acid binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF321P
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF321P
Gene Accession No. Q8N8H1
Gene Alias zinc finger protein 321, pseudogene; ZNF321; ZNF321P
Gene Symbols ZNF321P
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human ZNF321P.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 399669
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.