Learn More
Aviva Systems Biology Corp ZNF407 Antibody - middle region (ARP39306_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39306_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF407. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human ZNF407. Alternate names: FLJ13839, FLJ20307, KIAA1703ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-798