Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF418 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543629
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543629 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543629 Supplier Invitrogen™ Supplier No. PA543629
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: HRCEAWGNKL YDSSNRPHQN QYLGEKPYRS SVEEALFVKR CKFHVSEESS Sequence homology: Human: 100%.

ZNF418 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 16 C2H2-type zinc fingers and 1 KRAB domain. ZNF418 is a transcriptional repressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF418
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF418
Gene Accession No. Q8TF45
Gene Alias FLJ31551; KIAA1956; MGC138449; Zinc finger protein 418; ZNF418
Gene Symbols ZNF418
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ZNF418 (aa 110-159).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147686
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.