Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF488 Antibody (PCRP-ZNF488-2D8) [Alexa Fluor™Plus 405], R∓D Systems™
SDP

Catalog No. RD10083856
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10083856 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RD10083856 Supplier R&D Systems Supplier No. NBP327049AFP405

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

ZNF488 Monoclonal antibody specifically detects ZNF488 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry (Paraffin)

Specifications

Antigen ZNF488
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PCRP-ZNF488-2D8
Conjugate Alexa Fluor Plus 405
Formulation 50mM Sodium Borate
Gene Alias FDZF2Zinc finger protein FDZF2, zinc finger protein 230
Host Species Mouse
Immunogen Recombinant fragment (around aa170-307) of human ZNF488
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 118738
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.