Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF516 Antibody [Alexa Fluor Plus 680], R&D Systems™
SDP

Catalog No. RD10038978
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10038978 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RD10038978 Supplier R&D Systems Supplier No. NBP298464AFP680

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

ZNF516 Polyclonal antibody specifically detects ZNF516 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen ZNF516
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor Plus 680
Formulation 50mM Sodium Borate
Gene Alias HsT287, KIAA0222, zinc finger protein 516
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human ZNF516 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9658
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.