Learn More
Invitrogen™ ZNF521 Polyclonal Antibody

Description
ZNF521 Polyclonal Antibody for IHC (P), ICC/IF
Specifications
Specifications
| Antigen | ZNF521 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.11 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 30% glycerol and 0.05% sodium azide |
| Gene | ZNF521 |
| Gene Accession No. | Q6KAS7, Q96K83 |
| Gene Alias | B930086A16Rik; early hematopoietic zinc finger protein; Ecotropic viral integration site 3 protein; EHZF; Evi3; LIP3; LYST-interacting protein 3; Zfp521; Zinc finger protein 521; Znf521 |
| Gene Symbols | Zfp521, ZNF521 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.