Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF593 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501732400 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-2400 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-2400 Supplier Proteintech Group Inc Supplier No. 194261AP150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Negatively modulates the DNA binding activity of Oct-2 and therefore its transcriptional regulatory activity. Could act either by binding to DNA octamer or by interacting with Oct-2. May also be a modulator of other octamer-binding proteins.

Specifications

Antigen ZNF593
Applications Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ZNF593
Gene Accession No. O00488, Q9DB42
Gene Alias zinc finger protein 593, Zinc finger protein T86, ZNF593, ZT86
Gene Symbols ZNF593
Host Species Rabbit
Immunogen ZNF593 Fusion Protein Ag13758
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51042, 68040
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.