Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF606 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501735223 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5223 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5223 Supplier Proteintech Group Inc Supplier No. 261281AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May act as a transcriptional repressor.

Specifications

Antigen ZNF606
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.14 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ZNF606
Gene Accession No. Q8WXB4
Gene Alias KIAA1852, Zinc finger protein 328, zinc finger protein 606, ZNF328, ZNF606
Gene Symbols Zfp606, ZNF606
Host Species Rabbit
Immunogen ZNF606 Fusion Protein Ag23589
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 67370, 80095
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.