Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF622 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557699
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557699 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557699 Supplier Invitrogen™ Supplier No. PA557699
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MMNEKNLEKG LGVDSVDKDA MNAAIQQAIK AQPSMSPKKA PPAPAKEARN VVAVGTGGRG THDRDPSEKP PRLQWFEQQA KKLAKQQ Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 80%.

ZNF622 may behave as an activator of the bound transcription factor, MYBL2, and be involved in embryonic development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF622
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF622
Gene Accession No. Q969S3
Gene Alias 1110033B05Rik; D15Ertd806e; liver regeneration-related protein LRRG121; Zfp622; Zinc finger protein 622; zinc finger-like protein 9; Znf622; ZPR9
Gene Symbols ZNF622
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF622. Recombinant protein control fragment (Product #RP-96941).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90441
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.