Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF699 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501733484 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-3484 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-3484 Supplier Proteintech Group Inc Supplier No. 215621AP150UL

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

ZNF699 may be involved in transcriptional regulation.

Specifications

Antigen ZNF699
Applications Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ZNF699
Gene Accession No. Q32M78
Gene Alias hang, Hangover homolog, zinc finger protein 699, ZNF699
Gene Symbols ZNF699
Host Species Rabbit
Immunogen ZNF699 Fusion Protein Ag15990
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 374879
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.