Learn More
Invitrogen™ ZNF70 Monoclonal Antibody (1D8)

Description
Peptide Sequence: MEVPPATKFG ETFAFENRLE SQQGLFPGED LGDPFLQERG LEQMAVIYKE IPLGEQDEEN DDYEGNFSLC SSPVQHQSIP PGTRPQDDEL FGQTFLQKSD LSMCQIIHSE EPSPCDCAET DRGDSGPNAP HRTPQPAKPY ACRECGKAFS QSSHLLRHLV IHTGEKPYEC CECGKAFSQS SHLLRHQIRE CGKAFRQSSA LTQHQKIHTG KRPYECRECG KDFSRSSSLR KHERIHTGER PYQCKECGKS FNQSSGLSQH RKIHTLKKPH ECDLCGKAFC HRSHLIRHQR IHTGKKPYKC DECGKAFSQS SNLIEHRKTH TGEKPYKCQK CGKAFSQSSS LIEHQRCQCG KAFCHSSALI QHQRIHTGKK PYTCECGKAF RHRSALIEHY KTHTREKPYV CNLCGKSFRG SSHLIRHQKI HSGEKL.
Specifications
Specifications
| Antigen | ZNF70 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 1D8 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ZNF70 |
| Gene Accession No. | Q9UC06 |
| Gene Alias | Cos17; Zinc finger protein 70; zinc finger protein N27C7-1; ZNF70 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.