Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF703 Antibody [mFluor Violet 450 SE], R&D Systems™
SDP

Catalog No. NB797923
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB797923 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB797923 Supplier R&D Systems Supplier No. NBP232138MFV450

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

ZNF703 Polyclonal antibody specifically detects ZNF703 in Human samples. It is validated for Immunoprecipitation,Western Blot

Specifications

Antigen ZNF703
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Formulation 50mM Sodium Borate
Gene Alias FLJ14299, ZEPPO1, Zinc finger elbow-related proline domain protein 1, zinc finger protein 703, ZNF503L, ZPO1
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 100 to 150 of human Zinc Finger Protein 703 using the numbering given in entry NP_079345.1 (GeneID 80139).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80139
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.