Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF707 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521803
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521803 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521803 Supplier Invitrogen™ Supplier No. PA521803
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HepG2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The ZNF707 gene, a member of the zinc-finger protein (ZNF) family, encodes a transcription factor that can significantly influence various cellular processes by binding to DNA and regulating gene expression. Located on chromosome 4q32, zinc-finger proteins like ZNF707 are characterized by their zinc-finger motifs, which enable their interaction with specific DNA sequences. Mutations in ZNF707, such as rs4873815-TT, have been implicated in the tumorigenesis and prognosis of ovarian clear cell adenocarcinoma (Ov-CCA), highlighting its clinical significance in cancer research. Research also suggests that missense mutations in ZNF genes can alter protein-DNA interactions, potentially leading to cancer development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF707
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene ZNF707
Gene Accession No. Q96C28
Gene Alias Zinc finger protein 707; ZNF707
Gene Symbols ZNF707
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ZNF707. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286075
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.