Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF785 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541223
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541223 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541223 Supplier Invitrogen™ Supplier No. PA541223
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: RKTALEAHRW IHRSCSERRA WQQAVVGRSE PIPVLGGKDP PVHFRHFPDI Sequence homology: Dog: 79%; Human: 100%.

The function of ZNF785 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF785
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF785
Gene Accession No. A8K8V0
Gene Alias Zinc finger protein 785; zinc finger protein 785; LOW QUALITY PROTEIN: zinc finger protein 785; ZNF785
Gene Symbols ZNF785
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human ZNF785 (aa 351-400).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146540
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.