Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF8 Mouse anti-Human, Clone: OTI2B6, liquid, TrueMAB™
SDP

Catalog No. 501684360
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-4360 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-4360 Supplier OriGene Supplier No. TA811985
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Transcriptional repressor. May modulate BMP and TGF-beta signal transduction, through its interaction with SMAD proteins.

Specifications

Antigen ZNF8
Applications Western Blot
Classification Monoclonal
Clone OTI2B6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ZNF8
Gene Accession No. P17098
Gene Alias HF.18, Zfp128
Gene Symbols ZNF8
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 1-242 of human ZNF8 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7554
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.