Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF828 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501735304 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5304 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5304 Supplier Proteintech Group Inc Supplier No. 263151AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Required for proper alignment of chromosomes at metaphase and their accurate segregation during mitosis. Involved in the maintece of spindle microtubules attachment to the kinetochore during sister chromatid biorientation. May recruit CENPE and CENPF to the kinetochore.

Specifications

Antigen ZNF828
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene CHAMP1
Gene Accession No. Q96JM3
Gene Alias C13orf8, KIAA1802, zinc finger protein 828, ZNF828
Gene Symbols CHAMP1
Host Species Rabbit
Immunogen ZNF828 Fusion Protein Ag23698
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283489
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.