Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF830 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581132
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581132 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581132 Supplier Invitrogen™ Supplier No. PA581132
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human ZNF830.

Located on chromosome 17, this gene encodes for a coiled-coil domain containing 16 protein with unknown function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF830
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ZNF830
Gene Accession No. Q96NB3
Gene Alias 2410003C20Rik; AV301118; AW048207; CCDC16; coiled-coil domain containing 16; coiled-coil domain-containing protein 16; hypothetical protein MGC20398; MGC20398; Omcg1; orphan maintenance of genome 1; Ovus mutant candidate gene 1 protein; Zfp830; Zinc finger protein 830; Zinc finger protein 830-like protein; Znf830
Gene Symbols ZNF830
Host Species Rabbit
Immunogen E. coli-derived Human ZNF830 fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91603
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.