Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF839 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540619
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540619 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540619 Supplier Invitrogen™ Supplier No. PA540619
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: KMCQDYLSSS GLCSQETLEI NNDKVAESLG ITEFLRKKEI HPDNLGPKHL Sequence homology: Cow: 82%; Horse: 91%; Human: 100%.

ZNF839, also known as C14orf131, is an 811 amino acid protein that contains one C2H2-type zinc finger. The gene encoding ZNF839 localizes to chromosome 14 and, due to alternative splicing events, four ZNF839 isoforms are expressed. Chromosome 14 contains about 700 genes and 106 million base pairs and makes up about 3.5% of human cellular DNA. Chromosome 14 encodes the presinilin 1 (PSEN1) gene, which is one of the three key genes associated with the development of Alzheimer's disease. The SERPINA1 gene is located on chromosome 14 and when defective leads to the genetic disorder alpha1-antitrypsin deficiency. This disorder is characterized by severe lung complications and liver dysfunction. Notably, the immunoglobulin heavy chain locus is found on chromosome 14 and has been identified as a fusion with the chromosome 19 encoded protein BCL3 in the (14; 19) translocations found in a variety of B cell malignancies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF839
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF839
Gene Accession No. A8K0R7
Gene Alias C14orf131; renal carcinoma antigen NY-REN-50; Zinc finger protein 839; ZNF839
Gene Symbols ZNF839
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human C14ORF131.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55778
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.