Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNHIT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. p-7208026 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-1043 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-1043 Supplier Proteintech Group Inc Supplier No. 165951AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Seems to play a role in p53-mediated apoptosis induction.

Specifications

Antigen ZNHIT1
Applications Western Blot
Classification Polyclonal
Concentration 0.44 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ZNHIT1
Gene Accession No. O43257, Q8R331
Gene Alias CG1I, CGBP1, Cyclin G1 binding protein 1, p18 Hamlet, zinc finger, HIT type 1, ZNFN4A1, ZNHIT1
Gene Symbols Znhit1
Host Species Rabbit
Immunogen ZNHIT1 Fusion Protein Ag9741
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100360077, 10467, 70103
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.