Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNRF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555632
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555632 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555632 Supplier Invitrogen™ Supplier No. PA555632
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ASDSTYAHGN GYQETGGGHH RDGMLYLGSR ASLADALPLP IAPRWFSSHS GFKCPICSKS VASDEMEMHF I Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

E3 ubiquitin-protein ligase that mediates the ubiquitination of AKT1 and GLUL, thereby playing a role in neuron cells differentiation. Plays a role in the establishment and maintenance of neuronal transmission and plasticity. Regulates Schwann cells differentiation by mediating ubiquitination of GLUL. Promotes neurodegeneration by mediating 'Lys-48'-linked polyubiquitination and subsequent degradation of AKT1 in axons: degradation of AKT1 prevents AKT1-mediated phosphorylation of GSK3B, leading to GSK3B activation and phosphorylation of DPYSL2/CRMP2 followed by destabilization of microtubule assembly in axons (Probable).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNRF1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNRF1
Gene Accession No. Q8ND25
Gene Alias B830022L21Rik; E3 ubiquitin-protein ligase ZNRF1; MGC101991; nerve injury gene 283; nerve injury-induced gene 283 protein; Nin283; peripheral nerve injury protein nin283; ring finger protein 42; RING-type E3 ubiquitin transferase ZNRF1; Rnf42; zgc:77896; zinc and ring finger 1; zinc and ring finger 1, E3 ubiquitin protein ligase; zinc and ring finger protein 1; zinc ring finger protein 1; zinc/RING finger protein 1; ZNRF1; Zrfp1
Gene Symbols ZNRF1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNRF1. Recombinant protein control fragment (Product #RP-93636).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84937
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.