Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNRF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143891 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143891 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143891 Supplier Invitrogen™ Supplier No. PA5143891
  • $482.00
    $415.65 / Each of 1

    Save $66.35

    14% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human CACO-2 whole cell, human A549 whole cell, human Hacat whole cell. ICC/IF: U20S cell. Flow: PC-3 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

The ZNRF3 gene encodes a transmembrane E3 ubiquitin ligase that plays a critical role in regulating the Wnt signaling pathway, which is essential for various biological processes, including embryonic development and cell proliferation. ZNRF3 functions by negatively regulating this pathway through ubiquitination and degradation of Frizzled receptors, thereby suppressing the Wnt signaling. Located on chromosome 22, ZNRF3 acts as a tumor suppressor gene by inhibiting oncogenic Wnt signaling and has been implicated in the formation and progression of several cancers, particularly colorectal cancer. Moreover, ZNRF3 interacts with R-spondin proteins which are known to augment Wnt signaling by targeting ZNRF3 for degradation, thus facilitating cell differentiation and proliferation. Alterations in ZNRF3 expression or mutations can lead to aberrant Wnt signaling, contributing to tumorigenesis. Additionally, ZNRF3's role extends to its participation in lipid metabolism and maintenance of bone homeostasis. It has been studied as a potential therapeutic target due to its central involvement in these regulatory pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNRF3
Applications ELISA, Flow Cytometry, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene ZNRF3
Gene Accession No. Q9ULT6
Gene Alias BK747E2.3; CTA-292E10.6; E3 ubiquitin-protein ligase ZNRF3; Gm1167; KIAA1133; novel C3HC4 type Zinc finger (ring finger); rCG35969-like; RING finger protein 203; RING-type E3 ubiquitin transferase ZNRF3; RNF203; similar to ring finger protein 43; zinc and ring finger 3; zinc/RING finger protein 3; ZNRF3
Gene Symbols ZNRF3
Host Species Rabbit
Immunogen E.coli-derived human ZNRF3 recombinant protein (Position: R54-E913).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84133
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.