Learn More
Invitrogen™ ZO-1 Monoclonal Antibody (ZO1-1A12), Alexa Fluor™ 647
Shop All Thermo Scientific Products

Description
ZO-1 Monoclonal Antibody for ICC/IF
Specifications
Specifications
| Antigen | ZO-1 |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Clone | ZO1-1A12 |
| Concentration | 1 mg/mL |
| Conjugate | Alexa Fluor 647 |
| Formulation | PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4 |
| Gene | TJP1 |
| Gene Accession No. | O97758, Q07157 |
| Gene Alias | DKFZp686M05161; I79_005485; MGC133289; OTTHUMP00000174520; Tight junction protein 1; tight junction protein 1 (zona occludens 1); tight junction protein ZO-1; Tjp1; zo1; ZO-1; ZO-1 MDCK; zona occludens 1; zona occludens protein 1; zonula occludens 1; zonula occludens 1 protein; zonula occludens protein 1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.