Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZO2 Polyclonal Antibody, Huabio

Catalog No. 502544405
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4405 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4405 Supplier HUABIO RESEARCH INC Supplier No. R14022
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZO2 Polyclonal antibody specifically detects ZO2 in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen ZO2
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene TJP2
Gene Accession No. Q9UDY2, Q9Z0U1
Gene Alias C9DUPq21.11; DFNA51; DUP9q21.11; Friedreich ataxia region gene X104 (tight junction protein ZO-2); PFIC4; tight junction protein; tight junction protein 2; tight junction protein 2 (zona occludens 2); tight junction protein ZO-2; Tight junction protein ZO-2 (Zonula occludens 2 protein) (Zona occludens 2 protein) (Tight junction protein 2); Tjp2; X104; ZO2; ZO-2; zona occludens 2; zona occludens protein 2; Zonula occludens protein 2
Host Species Rabbit
Immunogen Synthetic peptide within C-terminal human ZO2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21873, 9414
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.