Learn More
BURKLE INC Jumbo sampler
Supplier:  BURKLE INC 53192150
Encompass_Preferred
The Multi jumbo sampler separates sample material using separators allowing multiple point samples to be taken from different layers of a bulk material These can be analyzed specifically or combined to form a mixed sample Autoclavable and flame retardant for minimal risk of contamination
Catalog No. 50-000-25875
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.