Learn More
Apexbio Technology LLC Zoniporide (hydrochloride) 241800-97-5 50mg
SDP
Supplier:  Apexbio Technology LLC C348050
Encompass_Preferred
Zoniporide (hydrochloride) (CAS 241800-97-5) is a potent and selective inhibitor of sodium-hydrogen exchanger isoform-1 (NHE-1) the primary regulator of intracellular pH in cardiac myocytes Zoniporide exhibits high selectivity for NHE-1 inhibiting the human isoform with an IC50 of 14 nM and demonstrating over 150-fold selectivity relative to other NHE isoforms In preclinical studies zoniporide reduced infarct size in isolated hearts and animal models of myocardial ischemia-reperfusion injury and attenuated NHE-1-mediated platelet swelling These properties make zoniporide a valuable tool for studying NHE-1 function in cardiovascular research and ischemia-reperfusion injury models
Catalog No. 50-000-89419
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.