Learn More
Abcam Zonisamide, Na+ and Ca2+ Channel Blocker, >99% Purity, 10 mM/mL
Description
- Purity: >99%
- Molecular Weight: 212.23 Da
- Mechanism: Blocks voltage-sensitive Na+ and T-type Ca2+ channels and stimulates BKCa channels
- Function: Broad spectrum antiepileptic; modulates GABA, glutamate, cholinergic and monoamine neurotransmission
- Additional properties: Exhibits free-radical scavenging properties
- Solubility: Soluble in 1 mL water to give specified mM/mL concentration
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Buffer | 100% Zonisamide |
| Shipping Condition | Ambient |
| Target Molecule | Na+ Channels, Ca2+ Channels, BKCa Channels, GABA, Glutamate, Cholinergic, Monoamine |
| Concentration | 10 mM |
| Content And Storage | Store at 4°C. |
| Formulation | 100% Zonisamide |
| Quantity | 10 mM |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.