Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ Zorb-IT™ Blacklite Nitrile Foam Coated Nylon Gloves

Catalog No. 19172197 Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
7
9
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-172-197 9
19-172-195 7
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-172-197 Supplier SHOWA™ Supplier No. 454009
This item is not returnable. View return policy

Sponge nitrile coating is absorbent and provides enhanced grip in wet or oily applications.

  • Nitrile foam coating is ideal in applications that require fit, feel, and touch sensitivity
  • Gloves are absorbent and enhance grip in wet or oily applications
  • Sanitized with antimicrobial bacteriostatic lining

Specifications

ANSI Abrasion Rating ANSI 4
Coated Area Palm
Construction Seamless Knit
Coating Material Nitrile Foam
Color Black
Exterior Finish No Texture
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size 9
For Use With (Application) Wet and oily conditions
Glove Cuff Style Knit
Series 4540
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.