Learn More
Market Lab Zorb Sheets, 3"""" x 3"""", 1000/pk
Supplier:  Market Lab 15246
Encompass_Preferred
Zorb and Super Barrier Zorb Sheets are specially designed with absorbent materials that help keep you and the patient safe. Both absorb and encapsulate potentially infectious spills, while preventing leakage and possible contamination of the handler.
Catalog No. NC1244727