Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101973
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101973 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101973 Supplier Invitrogen™ Supplier No. PA5101973
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ZP1.

The mammalian zona pellucida, which mediates species-specific sperm binding, induction of the acrosome reaction and prevents post-fertilization polyspermy, is composed of three to four glycoproteins, ZP1, ZP2, ZP3, and ZP4. ZP1 ensures the structural integrity of the zona pellucida.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ZP1
Gene Accession No. O54766, P60852, Q62005
Gene Alias epididymis luminal protein 163; HEL163; mgc87693; OOMD; OOMD1; Processed zona pellucida sperm-binding protein 1; Zona pellucida glycoprotein 1; zona pellucida glycoprotein 1 (sperm receptor); zona pellucida sperm-binding protein 1; ZP1; Zp-1; zp1 protein
Gene Symbols ZP1
Host Species Rabbit
Immunogen A synthesized peptide derived from human ZP1(Accession P60852), corresponding to amino acid residues I227-T277.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22786, 22917, 85271
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.