Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537086
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537086 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537086 Supplier Invitrogen™ Supplier No. PA537086
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 59 kDa. Purity is >95% by SDS-PAGE.

The mammalian zona pellucida is composed of four major glycoproteins, ZP1, ZP2, ZP3 and ZP4, which may act as sperm receptors. Two forms of porcine ZP4 peptides exist: one consisting of 128 amino acid residues and the other of 133 amino acid residues. These two peptides are identical, except the larger form contains an additional five amino acid sequence at its carboxy-terminal end. Both peptides have two potential N-linked glycosylation sites. The smaller peptide shares 39.1% identity with the amino-terminal region of mouse ZP2 polypeptide. Based on results from animal studies, ZP4 antigen is a promising candidate for the development of a contraceptive vaccine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZP4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide; pH 7.2
Gene ZP4
Gene Accession No. Q12836, Q8CH34
Gene Alias Processed zona pellucida sperm-binding protein 4; ZBP; zona pellucida 4; zona pellucida B glycoprotein; zona pellucida glycoprotein 4; Zona pellucida protein B; zona pellucida sperm-binding protein 4; ZP1; ZP4; Zp-4; ZPB
Gene Symbols ZP4
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 277-322 of human ZP4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282833, 57829
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.