Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZPLD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567625
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567625 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567625 Supplier Invitrogen™ Supplier No. PA567625
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Mouse samples.

ZPLD1 is a protein coding gene. Glycoprotein which is a component of the gelatinous extracellular matrix in the cupulae of the vestibular organ.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZPLD1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene ZPLD1
Gene Accession No. Q8BGZ8, Q8TCW7
Gene Alias 9430016A21Rik; Cupulin; zona pellucida like domain containing 1; zona pellucida-like domain containing 1; zona pellucida-like domain-containing protein 1; Zona pellucida-like domain-containing protein 1, secreted form; ZP domain-containing protein 1; ZPLD1
Gene Symbols ZPLD1
Host Species Rabbit
Immunogen Synthesized peptide derived from human ZPLD1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 131368, 239852
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.