Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZSCAN18 Monoclonal Antibody (OTI1F10), TrueMAB™, OriGene
SDP

Catalog No. 502539804
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-253-9804 30 μL
50-167-5792 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-253-9804 Supplier OriGene Supplier No. TA505284S
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZSCAN18 Monoclonal antibody specifically detects ZSCAN18 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen ZSCAN18
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1F10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene B3GNT2
Gene Accession No. Q9NY97
Gene Alias EG232875; RGD1560671; similar to hypothetical protein FLJ12895; zinc finger and SCAN domain containing 18; zinc finger and SCAN domain-containing protein 18; Zinc finger protein 447; ZNF447; ZSCAN18
Host Species Mouse
Immunogen Full length human recombinant protein of human B3GNT2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10678
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.