Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZSCAN18 Mouse anti-Human, Clone: UMAB113, liquid, UltraMAB™
SDP

Catalog No. 501684985
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-4985 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-4985 Supplier OriGene Supplier No. UM500080
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZNF447 contains 1 SCAN box domain and 2 C2H2-type zinc fingers. It belongs to the kruppel C2H2-type zinc-finger protein family and may be involved in transcriptional regulation.

Specifications

Antigen ZSCAN18
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone UMAB113
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ZSCAN18
Gene Accession No. Q8TBC5
Gene Alias ZNF447
Gene Symbols ZSCAN18
Host Species Mouse
Immunogen Full length human recombit protein of human ZSCAN18 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65982
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.