Learn More
Description
Specifications
Specifications
| Gene ID (Entrez) | 201516 |
| Species | Human |
| Product Line | Novus |
| Content And Storage | Store at -20 C. Avoid freeze-thaw cycles. |
| Formulation | PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NBP1-77120. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | FLJ35105, MGC126787, zinc finger and SCAN domain containing 4, Zinc finger protein 494 mgC126789, ZNF494zinc finger and SCAN domain-containing protein 4 |
| Gene Symbol | ZSCAN4 |
| Product Type | ZSCAN4 |
| Quantity | 0.05 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
