Learn More
ATCC Zygosaccharomyces Parabailii Suh et al. - Cryopreserved QC yeast strain (ATCC MYA-4549)
Supplier:  ATCC MYA4549
Encompass
Zygosaccharomyces parabailii is a cryopreserved yeast strain supplied as a frozen ampoule containing viable cells suspended in cryoprotectant. The strain is intended for use as a quality control organism for yeast identification systems and related microbiological QC applications. Store at -80°C or colder and handle according to institutional biosafety procedures.
  • Supplied as a frozen ampoule containing viable cells in cryoprotectant.
  • Intended for quality control of yeast identification assays.
  • Suitable for preparation of control panels and performance verification.
  • Requires storage at -80°C or colder to maintain viability.
  • Biosafety level 1 organism; handle according to institutional safety procedures.
Catalog No. NC3440910