Learn More
Cytiva (Formerly Pall Lab) PTFE Membrane Disc Filter - 0.2 m, 47 mm (100/pkg)
Supplier:  Cytiva 66143
Encompass_Preferred
Strong, Chemically Resistant Membranes for Air Monitoring and Sampling in Aggressive EnvironmentsLow chemical background permits highly sensitive, interference-free determinations.Ensures accurate gravimetric determinations with low tare mass.Ideal for filtration of gas and/or organic solvents. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3281
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.