Learn More
ZymeTx ZstatFlu™ Test for Influenza Types A and B
Description
- Test results within 20 minutes
- CLIA Complexity: Waived
- An endogenous viral-encoded enzyme assay for the qualitative determination of influenza Types A and B from throat swab specimens
- Test requires no special handling or reading instruments
- Simply obtain specimen, combine with diluent, incubate, add stop reagent and read results
Specifications
Specifications
| Detectable Analytes | Influenza A and B |
| Type | Collection Swab |
| Quantity | 50 Case |