Learn More
Research Products International Corp Zymo-Seq Cell Free DNA WGBS
SDP
Supplier:  Research Products International Corp ZD5463
Encompass_Preferred
Zymo-Seq Cell Free DNA WGBS Library Kit provides an optimized and reliable workflow for whole genome bisulfite sequencing (WGBS) library preparation from cell-free DNA (cfDNA). This all-inclusive kit features a straightforward procedure capable of preparing high-quality methyl-seq libraries from as little as 5 ng of cfDNA. The process is completed in three basic steps: bisulfite conversion, direct adapter ligation, and index PCR amplification. The initial bisulfite treatment is gentle on cfDNA yet effective in converting any unmodified cytosines into uracil. Next, the innovative splinted adapters capture and directly ligate the Illumina-compatible adapters onto any size DNA fragment, allowing for sequencing of nicked, damaged, and short DNA fragments that may have otherwise been discarded with traditional library prep methods.
Catalog No. 50-238-8382
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.