Learn More
Research Products International Corp Zymo Seq miRNA Library Kit
SDP
Supplier:  Research Products International Corp ZR3007
Encompass_Preferred
The Zymo-Seq miRNA Library Kit is an innovative small RNA sequencing library prep kit that uses a single-adapter and circularization strategy to reduce ligation bias and provide accurate small RNA profiling from cell-free RNA (cfRNA) and total RNA. This simple, streamlined workflow minimizes hands-on time and uses gel-free removal of adapter dimers to generate stranded small RNA libraries from plasma, biofluids, cells, tissues, and more.
Catalog No. 50-238-8421
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.