Learn More
Research Products International Corp Zymo Spin V Col Uncapped
SDP
Supplier:  Research Products International Corp ZC101250
Encompass_Preferred
The versatile Zymo-Spin V Fast-Spin column can be used either in microcentrifuges, centrifuges, or on vacuum manifolds for the purification of DNA and/or RNA. This column features a luer-lock top allowing it to be easily attached to a syringe. The Zymo-Spin V features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 100 µg DNA or RNA in ≥ 150 µl eluate. Capacity is 800 µl.
Catalog No. 50-238-8444
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.