Learn More
Research Products International Corp Zymo-Spin V-PS Col Ambly
SDP
Supplier:  Research Products International Corp ZC10835
Encompass_Preferred
Available as a refill for the ZymoPURE II Plasmid Midiprep and ZymoPURE Express Plasmid Midiprep Kits. The versatile Zymo-Spin V-PS with 15 ml and 50 ml reservoirs can be used in centrifuges or on vacuum manifolds for the purification of plasmid DNA. The Zymo-Spin V-PS features durable polypropylene construction and a unique silica-based matrix that allows for purification of up to 1.2 mg of plasmid DNA in 150 l eluate when used in combination with ZymoPURE Plasmid Prep buffers. Capacity with reservoir assembly is 65 ml.
Catalog No. 50-238-8394
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.