Learn More
United States Biological Corporation Zymolyase 100T Concentrate, 1 mL
Catalog No. NC1117530 Shop All United States Biological Corporation Products
Encompass_Preferred
Zymolyase, produced by a submerged culture of Arthrobacter luteus, is an enzyme preparation which effectively lyses cell walls of viable yeast cells. An essential enzyme responsible for lysis of viable yeast cells in this preparation is b-1, 3-glucan laminaripentaohydrolase.
Catalog No. NC1117530 Supplier United States Biological Corporation Supplier No. Z10051ML
Zymolyase, produced by a submerged culture of Arthrobacter luteus, is an enzyme preparation which effectively lyses cell walls of viable yeast cells. An essential enzyme responsible for lysis of viable yeast cells in this preparation is b-1, 3-glucan laminaripentaohydrolase.